KEY388193171 Key Organics / BIONET

(S)-3-(dimethylamino)-1-phenylpropan-1-ol; oxalic acid

  • MDL number MFCD23136037
  • Cas Number : 873436-53-4
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357493

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4576_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart