KEY388193183 Key Organics / BIONET

(1S)-1-amino-1-phenylbut-3-yne hydrochloride

  • MDL number MFCD23136027
  • Cas Number : 1423802-96-3
  • Molecular Weight : 181.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108192

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-2962_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart