KEY388193192 Key Organics / BIONET

5-bromo-6-(trifluoromethyl)-1H-indole-3-carbaldehyde

  • MDL number MFCD23136030
  • Cas Number : 1423757-77-0
  • Molecular Weight : 292.055

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 272 - 275
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357451

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AH-0711_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart