KEY388193195 Key Organics / BIONET

2-fluoro-5-[(4-oxo-3,4-dihydrophthalazin-1-yl)methyl]benzoic acid

  • MDL number MFCD14636678
  • Cas Number : 763114-26-7
  • Molecular Weight : 298.273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357496

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4579_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart