KEY388193196 Key Organics / BIONET

1,3-diethyl 2-(3,6-dichloro-5-nitropyridin-2-yl)propanedioate

  • MDL number MFCD23136038
  • Cas Number : 136888-77-2
  • Molecular Weight : 351.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357497

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4580_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart