KEY388193197 Key Organics / BIONET

18-(benzyloxy)-18-oxooctadecanoic acid

  • MDL number MFCD09991736
  • Cas Number : 189625-51-2
  • Molecular Weight : 404.591

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357498

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4581_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart