KEY388193198 Key Organics / BIONET

1,3-dichloro-7-methoxyisoquinoline

  • MDL number MFCD20441180
  • Cas Number : 24623-40-3
  • Molecular Weight : 228.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166 - 169
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357452

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AH-0722_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart