KEY388193200 Key Organics / BIONET

ethyl 4-(1-methyl-5-nitro-1H-1,3-benzodiazol-2-yl)butanoate

  • MDL number MFCD09840985
  • Cas Number : 3543-72-4
  • Molecular Weight : 291.307

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357501

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4584_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart