KEY388193202 Key Organics / BIONET

3,4-dihydro-2H-spiro[naphthalene-1,4'-piperidine] hydrochloride

  • MDL number MFCD02178985
  • Cas Number : 95195-98-5
  • Molecular Weight : 237.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357503

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4586_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart