KEY388193203 Key Organics / BIONET

6-fluoro-3-nitroimidazo[1,2-a]pyridine

  • MDL number MFCD20483121
  • Cas Number : 1235993-30-2
  • Molecular Weight : 181.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 169 - 170
Iupac Name 6-fluoro-3-nitroimidazo[1,2-a]pyridine
Inchi Code 1S/C7H4FN3O2/c8-5-1-2-6-9-3-7(11(12)13)10(6)4-5/h1-4H
Inchi Key WTEGDZZHPUPJCU-UHFFFAOYSA-N

Description

Melting Point 169 - 170
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357456

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BH-0703_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart