KEY388711412 Key Organics / BIONET

6-amino-3H,4H-pyrido[2,3-b]pyrazin-3-one

  • MDL number MFCD20697305
  • Cas Number : 874493-61-5
  • Molecular Weight : 162.152

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357560

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4599_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart