KEY388711414 Key Organics / BIONET

4,6-dichloro-N-methoxy-N-methylpyrimidine-5-carboxamide

  • MDL number MFCD23697509
  • Cas Number : 1426904-70-2
  • Molecular Weight : 236.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357563

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4602_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart