KEY388711418 Key Organics / BIONET

2-oxo-5-(pyridin-4-yl)-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD23697510
  • Cas Number : 62749-61-5
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357567

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4606_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart