KEY388711419 Key Organics / BIONET

5-(2-fluorophenyl)-2-methyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD08275134
  • Cas Number : 522646-43-1
  • Molecular Weight : 237.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(2-fluorophenyl)-2-methyl-1,3-thiazole-4-carboxylic acid
Inchi Code 1S/C11H8FNO2S/c1-6-13-9(11(14)15)10(16-6)7-4-2-3-5-8(7)12/h2-5H,1H3,(H,14,15)
Inchi Key ODCOWPJBSKLCIC-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/357569

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4608_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart