KEY390383044 Key Organics / BIONET

1',6'-dihydro-[2,3'-bipyridine]-6'-one

  • MDL number MFCD11113414
  • Cas Number : 381233-78-9
  • Molecular Weight : 172.187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152
Purity 97
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/358449

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/CH-0026_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart