KEY390383067 Key Organics / BIONET

2-(chloromethyl)-5-methyl-1,3,4-oxadiazole

  • MDL number MFCD06655111
  • Cas Number : 3914-42-9
  • Molecular Weight : 132.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Light sensitive
Physical Form Colourless to Dark Red Liquid
Shipping Temperature Light sensitive
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/358547

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P410
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4614_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart