KEY394378956 Key Organics / BIONET

tert-butyl 4-(trifluoromethanesulfonyloxy)-1,2,3,6-tetrahydropyridine-1-carboxylate

  • MDL number MFCD09997858
  • Cas Number : 138647-49-1
  • Molecular Weight : 331.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp LMS
Iupac Name tert-butyl 4-{[(trifluoromethyl)sulfonyl]oxy}-3,6-dihydro-1(2H)-pyridinecarboxylate
Inchi Code 1S/C11H16F3NO5S/c1-10(2,3)19-9(16)15-6-4-8(5-7-15)20-21(17,18)11(12,13)14/h4H,5-7H2,1-3H3
Inchi Key WUBVEMGCQRSBBT-UHFFFAOYSA-N

Description

Melting Point LMS
Purity 95%
Storage temp. Store under N2
Physical Form Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359402

Safety Information

signalword Warning
pcodes P422, P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DH-0724_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart