KEY394378969 Key Organics / BIONET

6-chloro-N-(2-cyclopropylethyl)pyridazine-3-carboxamide

  • MDL number MFCD21255380
  • Cas Number : 840488-85-9
  • Molecular Weight : 225.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359386

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DH-0024_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart