KEY394427565 Key Organics / BIONET

3-cyclopropyl-1H-pyrazole

  • MDL number MFCD04970938
  • Cas Number : 100114-57-6
  • Molecular Weight : 108.144

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 97
Storage temp. RT
Physical Form Off-white to Dark Yellow Liquid,Low Melting Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359681

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FH-0702_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart