KEY394427568 Key Organics / BIONET

2-(4-bromophenyl)-2-{[(tert-butoxy)carbonyl]amino}acetic acid

  • MDL number MFCD04115515
  • Cas Number : 917925-71-4
  • Molecular Weight : 330.178

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125 - 128
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359684

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FH-0710_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart