KEY394427572 Key Organics / BIONET

4,6-dichloro-5-(difluoromethyl)pyrimidine

  • MDL number MFCD25368744
  • Cas Number : 1443290-45-6
  • Molecular Weight : 198.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,6-dichloro-5-(difluoromethyl)pyrimidine
Inchi Code 1S/C5H2Cl2F2N2/c6-3-2(5(8)9)4(7)11-1-10-3/h1,5H
Inchi Key PKVJMACXCLXTKX-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store under N2
Physical Form Liquid,Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359688

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P402 + P404,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/FH-0725_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart