KEY394427578 Key Organics / BIONET

(2S,3S)-3-[(2-chlorophenyl)carbamoyl]-2,3-dihydroxypropanoic acid

  • MDL number MFCD25368745
  • Cas Number : 17447-41-5
  • Molecular Weight : 259.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359695

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4621_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart