KEY394427579 Key Organics / BIONET

tert-butyl 5-{[(benzyloxy)carbonyl]amino}pentanoate

  • MDL number MFCD08275837
  • Cas Number : 63983-88-0
  • Molecular Weight : 307.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359696

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4622_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart