KEY402333658 Key Organics / BIONET

3-(4-methoxyphenyl)-3H-1lambda4,2,3-oxadiazol-1-ylium-5-olate

  • MDL number MFCD25948962
  • Cas Number : 3815-80-3
  • Molecular Weight : 192.174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Solid
Purity 90
Storage temp. Store under N2 at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359930

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-1012_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart