KEY402333659 Key Organics / BIONET

4-iodo-3-(4-methoxyphenyl)-3H-1lambda4,2,3-oxadiazol-1-ylium-5-olate

  • MDL number MFCD25948963
  • Cas Number : 201282-63-5
  • Molecular Weight : 318.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Solid
Purity 95
Storage temp. Store under N2 at 4
Physical Form Off-white to Brown Solid
Shipping Temperature Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359931

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-1013_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart