KEY402333660 Key Organics / BIONET

3-(4-methoxyphenyl)-4-(trifluoromethyl)-3H-1lambda4,2,3-oxadiazol-1-ylium-5-olate

  • MDL number MFCD25948964
  • Cas Number : 1429485-53-9
  • Molecular Weight : 260.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Solid
Purity 95
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359932

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-1014_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart