KEY402333662 Key Organics / BIONET

potassium trifluoro[3-(4-methoxyphenyl)-1H-pyrazol-5-yl]boranuide

  • MDL number MFCD25948966
  • Cas Number : 1402242-82-3
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Solid
Purity 95
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359943

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-1027_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart