KEY402333669 Key Organics / BIONET

3-(difluoromethyl)-2-fluoro-4-iodopyridine

  • MDL number MFCD25479865
  • Cas Number : 1446786-38-4
  • Molecular Weight : 272.997

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Low melting solid
Purity 97
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359957

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FH-0736_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart