KEY402333671 Key Organics / BIONET

10,12-dioxo-5-(trifluoromethyl)-1lambda5,8,11,13-tetraazatetracyclo[7.6.1.0^{2,7}.0^{13,16}]hexadeca-1(16),2,4,6,8-pentaen-1-ylium chloride

  • MDL number MFCD25948722
  • Cas Number : 353245-38-2
  • Molecular Weight : 344.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Solid
Purity 95
Storage temp. Store under N2 at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359933

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-1015_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart