KEY402333674 Key Organics / BIONET

2-[1-methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]ethan-1-ol

  • MDL number MFCD25901663
  • Cas Number : 1446786-29-3
  • Molecular Weight : 194.157

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359936

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-1018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart