KEY402333679 Key Organics / BIONET

tert-butyl N-{[1-methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]methyl}carbamate

  • MDL number MFCD25948724
  • Cas Number : 1446786-36-2
  • Molecular Weight : 279.263

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 90
Storage temp. Store under N2 at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359941

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-1023_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart