KEY402333682 Key Organics / BIONET

methyl 2'-formyl-[1,1'-biphenyl]-4-carboxylate

  • MDL number MFCD04039121
  • Cas Number : 205823-24-1
  • Molecular Weight : 240.258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45 - 48
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3685

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Z-0710_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart