KEY402333683 Key Organics / BIONET

1-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trimethylsilyl)-1H-1,2,3-triazole

  • MDL number MFCD18383356
  • Cas Number : 1130489-19-8
  • Molecular Weight : 357.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Solid
Purity 85
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359926

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-1008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart