KEY402333693 Key Organics / BIONET

3-(1-benzylpiperidin-4-yl)-3,4-dihydro-1H-2lambda6,1,3-benzothiadiazine-2,2-dione

  • MDL number MFCD25948726
  • Cas Number : 79099-04-0
  • Molecular Weight : 357.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 99
Purity 97
Storage temp. RT
Physical Form Off-white to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359956

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/FH-0733_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart