KEY402333694 Key Organics / BIONET

potassium trifluoro(hex-1-yn-1-yl)boranuide

  • MDL number MFCD11052661
  • Cas Number : 244301-59-5
  • Molecular Weight : 188.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Solid
Purity 95
Storage temp. Store under N2 at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359921

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-1003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart