KEY402333705 Key Organics / BIONET

(2S,3S)-3-amino-2-hydroxy-4-phenylbutanoic acid

  • MDL number MFCD03788072
  • Cas Number : 62023-62-5
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108226

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3011_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart