KEY402333708 Key Organics / BIONET

5-chloro-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD11846318
  • Cas Number : 1240725-66-9
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/359969

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4641_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart