KEY407050945 Key Organics / BIONET

(3S,4S,5S,7S)-6-methyl-2-oxa-6-azatricyclo[3.3.1.0^{3,7}]nonan-4-ol

  • MDL number MFCD07783424
  • Cas Number : 487-27-4
  • Molecular Weight : 155.197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362596

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2013_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart