KEY407050959 Key Organics / BIONET

7-benzyl-4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD16660858
  • Cas Number : 1244855-59-1
  • Molecular Weight : 369.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362602

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart