KEY407050963 Key Organics / BIONET

3-bromo-9-ethyl-9H-carbazole

  • MDL number MFCD00092392
  • Cas Number : 57102-97-3
  • Molecular Weight : 274.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-9-ethyl-9H-carbazole
Inchi Code 1S/C14H12BrN/c1-2-16-13-6-4-3-5-11(13)12-9-10(15)7-8-14(12)16/h3-9H,2H2,1H3
Inchi Key PPHYYUDMDADQPF-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362606

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2033_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart