KEY407050976 Key Organics / BIONET

6-iodo-9H-purin-2-amine

  • MDL number MFCD08706375
  • Cas Number : 19690-23-4
  • Molecular Weight : 261.026

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362619

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart