KEY407050990 Key Organics / BIONET

methyl 4'-bromo-[1,1'-biphenyl]-3-carboxylate

  • MDL number MFCD05980426
  • Cas Number : 149506-25-2
  • Molecular Weight : 291.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4'-bromo[1,1'-biphenyl]-3-carboxylate
Inchi Code 1S/C14H11BrO2/c1-17-14(16)12-4-2-3-11(9-12)10-5-7-13(15)8-6-10/h2-9H,1H3
Inchi Key UTOKFKAMZICCBU-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362633

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2073_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart