KEY407050993 Key Organics / BIONET

3'-bromo-[1,1'-biphenyl]-4-carboxylic acid

  • MDL number MFCD08701660
  • Cas Number : 5737-83-7
  • Molecular Weight : 277.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3'-bromo[1,1'-biphenyl]-4-carboxylic acid
Inchi Code 1S/C13H9BrO2/c14-12-3-1-2-11(8-12)9-4-6-10(7-5-9)13(15)16/h1-8H,(H,15,16)
Inchi Key YZCQIXZSVVFEJE-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362636

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2081_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart