KEY407051026 Key Organics / BIONET

3'-bromo-[1,1'-biphenyl]-4-carbaldehyde

  • MDL number MFCD05979167
  • Cas Number : 400749-87-3
  • Molecular Weight : 261.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3'-bromo[1,1'-biphenyl]-4-carbaldehyde
Inchi Code 1S/C13H9BrO/c14-13-3-1-2-12(8-13)11-6-4-10(9-15)5-7-11/h1-9H
Inchi Key VVFSRYVBFDTNPL-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362669

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2146_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart