KEY407051029 Key Organics / BIONET

{[1,1'-biphenyl]-3-yl}methanamine hydrochloride

  • MDL number MFCD04117754
  • Cas Number : 870837-46-0
  • Molecular Weight : 219.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362672

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2150_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart