KEY407051031 Key Organics / BIONET

[3'-(trifluoromethyl)-[1,1'-biphenyl]-3-yl]methanol

  • MDL number MFCD05980218
  • Cas Number : 773872-61-0
  • Molecular Weight : 252.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3'-(trifluoromethyl)[1,1'-biphenyl]-3-yl]methanol
Inchi Code 1S/C14H11F3O/c15-14(16,17)13-6-2-5-12(8-13)11-4-1-3-10(7-11)9-18/h1-8,18H,9H2
Inchi Key IROMTEIILLDKHZ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362674

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2157_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart