KEY407051037 Key Organics / BIONET

(4-methoxyphenyl)({[(4-methoxyphenyl)dimethylsilyl]oxy})dimethylsilane

  • MDL number MFCD09266156
  • Cas Number : 122571-17-9
  • Molecular Weight : 346.573

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362680

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2192_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart