KEY407051055 Key Organics / BIONET

[2-({[1,1'-biphenyl]-4-yl}dimethylsilyl)phenyl]methanol

  • MDL number MFCD16251561
  • Cas Number : 1217863-72-3
  • Molecular Weight : 318.491

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362698

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2230_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart