KEY407051056 Key Organics / BIONET

(2-{dimethyl[4-(morpholine-4-carbonyl)phenyl]silyl}phenyl)methanol

  • MDL number MFCD16293906
  • Cas Number : 1244855-81-9
  • Molecular Weight : 355.509

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362699

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2232_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart