KEY407051058 Key Organics / BIONET

{2-[dimethyl(2,4,6-trimethylphenyl)silyl]phenyl}methanol

  • MDL number MFCD18643352
  • Cas Number : 947515-76-6
  • Molecular Weight : 284.474

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/362701

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2236_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart